Learn More
Biolegend Purified anti-HMGB1
Catalog No. 5000064470
Encompass_Preferred
Purified anti-HMGB1 [A15164F] Isotype Mouse IgG2a Reactivity Human Mouse Rat Apps WB ICC ICFC Size 25 g
Catalog No. 50-000-64470 Supplier Biolegend Supplier No. 641251
Purified anti-HMGB1 [A15164F] Isotype Mouse IgG2a Reactivity Human Mouse Rat Apps WB ICC ICFC Size 25 g