Learn More
Biolegend Purified anti-human CD11b (Maxpar Ready)
Supplier:  Biolegend 301337
Encompass_Preferred
Purified anti-human CD11b (Maxpar Ready) [ICRF44]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: FC, CyTOF; Size: 100 ug
Catalog No. NC1823996