Learn More
Biolegend Purified anti-human CD369 (Dectin-1/CLEC7A)
Supplier:  Biolegend 355402
Encompass_Preferred
Purified anti-human CD369 (Dectin-1/CLEC7A) [15E2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC, IHC-F; Size: 100 ug
Catalog No. NC1866704