Learn More
Biolegend Purified anti-human CD43
Catalog No. 5000064559
Encompass_Preferred
Purified anti-human CD43 [MT1] Isotype Mouse IgG1 Reactivity Human Apps FC IHC-P Size 100 g
Catalog No. 50-000-64559 Supplier Biolegend Supplier No. 314311
Purified anti-human CD43 [MT1] Isotype Mouse IgG1 Reactivity Human Apps FC IHC-P Size 100 g