Learn More
Biolegend Purified anti-human CD69 (Maxpar Ready)
Supplier:  Biolegend 310939
Encompass_Preferred
Purified anti-human CD69 (Maxpar Ready) [FN50]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC, CyTOF; Size: 100 ug
Catalog No. NC1217739