Learn More
Biolegend Purified anti-human FcRL6
Supplier:  Biolegend 373902
Encompass_Preferred
Purified anti-human FcRL6 [7B7/FcRL6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. NC1357132