Learn More
Biolegend Purified anti-Jun Phospho (Ser73) [A19012A/R/C]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, ICC; Size: 100 ug
Supplier:  Biolegend 605552
Encompass_Preferred
Purified anti-Jun Phospho (Ser73) [A19012A/R/C]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, ICC; Size: 100 ug
Catalog No. 50-237-6946