Learn More
Biolegend Purified anti-Keratin 14
Supplier:  Biolegend 905304
Encompass_Preferred
Purified anti-Keratin 14 [Poly19053]; Isotype: Rabbit Polyclonal; Reactivity: Human, Mouse, Rat; Cross-Reactivity: Dog, Primate; Apps: IHC-P, ICC, WB; Size: 100 ug
Catalog No. NC1300287