Learn More
Biolegend Purified anti-MLKL
Supplier:  Biolegend 666802
Encompass_Preferred
Purified anti-MLKL [W17104A]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB; Size: 100 ug
Catalog No. 50-208-7251