Learn More
Biolegend Purified anti-MOB1A
Supplier:  Biolegend 946701
Encompass_Preferred
Purified anti-MOB1A [W20004B]; Isotype: Rat IgG2b, κ; Reactivity: Human, Mouse; Apps: WB, IP; Size: 25 μg
Catalog No. 50-272-1595