Learn More
Biolegend Purified anti-mouse CD198 (CCR8)
Supplier:  Biolegend 150302
Encompass_Preferred
Purified anti-mouse CD198 (CCR8) [SA214G2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1106649