Learn More
Biolegend Purified anti-mouse CD24 (Maxpar Ready)
Supplier:  Biolegend 101829
Encompass_Preferred
Purified anti-mouse CD24 (Maxpar Ready) [M1/69]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, CyTOF; Size: 100 ug
Catalog No. NC1263666