Learn More
Biolegend Purified anti-mouse CD301a MGL
Catalog No. 502722650
Encompass_Preferred
Purified anti-mouse CD301a (MGL1) [LOM-8.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F, IP; Size: 100 μg
Catalog No. 50-272-2650 Supplier Biolegend Supplier No. 145602
Add to Cart
Add to Cart
Purified anti-mouse CD301a (MGL1) [LOM-8.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F, IP; Size: 100 μg