Learn More
Biolegend Purified anti-mouse I-Ad Recom
Catalog No. 502865953
Encompass_Preferred
Purified anti-mouse I-Ad Recombinant [39-10-8.RecM1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Catalog No. 50-286-5953 Supplier Biolegend Supplier No. 166902
Purified anti-mouse I-Ad Recombinant [39-10-8.RecM1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg