Learn More
Biolegend Purified anti-mouse PD-1H (VISTA)
Supplier:  Biolegend 143702
Encompass_Preferred
Purified anti-mouse PD-1H (VISTA) [MH5A]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC-F, WB; Size: 100 ug
Catalog No. NC1490985