Learn More
Biolegend Purified anti-mouse Siglec-G [SH1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Supplier:  Biolegend 163302
Encompass_Preferred
Purified anti-mouse Siglec-G [SH1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-237-8613