Learn More
Biolegend Purified anti-mTOR Phospho (Ser2448)
Supplier:  Biolegend 610302
Encompass_Preferred
Purified anti-mTOR Phospho (Ser2448) [A17024A]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, ICC; Size: 100 ug
Catalog No. 50-208-6687