Learn More
Biolegend Purified anti-NeuroD1
Catalog No. 502721451
Encompass_Preferred
Purified anti-NeuroD1 [W22035C]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB, ICC; Size: 100 μg
Catalog No. 50-272-1451 Supplier Biolegend Supplier No. 633452
Purified anti-NeuroD1 [W22035C]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB, ICC; Size: 100 μg