Learn More
Biolegend Purified (azide-free) anti-Neurofilament H (NF-H), Nonphosphorylated
Supplier:  Biolegend 801702
Encompass_Preferred
Purified (azide-free) anti-Neurofilament H (NF-H), Nonphosphorylated [SMI 32]; Isotype: Mouse IgG1; Reactivity: Human, Mouse, Rat, Other mammalian; Apps: IHC-P, WB, Array Tomography, ICC; Size: 25 uL
Catalog No. NC1490242