Learn More
Biolegend Purified anti-Neurofilament H
Catalog No. 502721663
Encompass_Preferred
Purified anti-Neurofilament H (NF-H), Phosphorylated [SMI 34]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB; Size: 25 μg
Catalog No. 50-272-1663 Supplier Biolegend Supplier No. 835504
Purified anti-Neurofilament H (NF-H), Phosphorylated [SMI 34]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB; Size: 25 μg