Learn More
Biolegend Purified anti-NMDAR1
Supplier:  Biolegend 828201
Encompass_Preferred
Purified anti-NMDAR1 [R1JHL]; Isotype: Mouse IgG1, κ; Reactivity: Rat, Mouse; Apps: WB; Size: 15 ug
Catalog No. 50-207-4252