Learn More
Biolegend Purified (azide-free) anti-NrCAM
Supplier:  Biolegend 821601
Encompass_Preferred
Purified (azide-free) anti-NrCAM [N343/26]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: WB, ICC, IHC-P; Size: 100 uL
Catalog No. NC1926276