Learn More
Biolegend Purified anti-Olig2
Catalog No. 502721814
Encompass_Preferred
Purified anti-Olig2 [W21023A]; Isotype: Rat IgG2b, κ; Reactivity: Human, Rat; Apps: IHC-P, ICC; Size: 25 μg
Catalog No. 50-272-1814 Supplier Biolegend Supplier No. 615951
Purified anti-Olig2 [W21023A]; Isotype: Rat IgG2b, κ; Reactivity: Human, Rat; Apps: IHC-P, ICC; Size: 25 μg