Learn More
Biolegend Purified anti-P-Cadherin
Supplier:  Biolegend 948402
Encompass_Preferred
Purified anti-P-Cadherin [12H6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IP; Size: 100 μg
Catalog No. 50-272-1578