Learn More
Biolegend Purified anti-P-Cadherin
Catalog No. 502721579
Encompass_Preferred
Purified anti-P-Cadherin [12H6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IP; Size: 25 μg
Catalog No. 50-272-1579 Supplier Biolegend Supplier No. 948401
Purified anti-P-Cadherin [12H6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IP; Size: 25 μg