Learn More
Biolegend Purified anti-P2RY12
Supplier:  Biolegend 848001
Encompass_Preferred
Purified anti-P2RY12 [S16007D]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: IHC-P, FC; Size: 25 ug
Catalog No. NC1380969