Learn More
Biolegend Purified anti-Pan-Shank
Supplier:  Biolegend 851901
Encompass_Preferred
Purified anti-Pan-Shank [N23B/49]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: WB, IHC-P, ICC; Size: 25 ug
Catalog No. 50-207-4657