Learn More
Biolegend Purified anti-Pax-6
Supplier:  Biolegend 901301
Encompass_Preferred
Purified anti-Pax-6 [Poly19013]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human, Mouse, Rat; Apps: WB, IHC-P, IHC-F, ICC; Size: 100 uL
Catalog No. 50-103-0098