Learn More
Biolegend Purified anti-PDPK1 Phospho (Ser241)
Supplier:  Biolegend 939602
Encompass_Preferred
Purified anti-PDPK1 Phospho (Ser241) [A19011A]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB; Size: 100 ug
Catalog No. 50-207-5407