Learn More
Biolegend Purified anti-Phosphothreonine [M380A]; Isotype: Mouse IgG1, ; Reactivity: Human, Mouse, Rat; Apps: WB, ICC, IP; Size: 100 ug
Supplier:  Biolegend 943002
Encompass_Preferred
Purified anti-Phosphothreonine [M380A]; Isotype: Mouse IgG1, ; Reactivity: Human, Mouse, Rat; Apps: WB, ICC, IP; Size: 100 ug
Catalog No. 50-207-5448