Learn More
Biolegend Purified (azide-free) anti-PPP2R4 (PTPA)
Supplier:  Biolegend 824401
Encompass_Preferred
Purified (azide-free) anti-PPP2R4 (PTPA) [5G3]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse; Apps: IHC-P, WB; Size: 100 uL
Catalog No. 50-207-4215