Learn More
Biolegend Purified (azide-free) anti-PSD95
Supplier:  Biolegend 810302
Encompass_Preferred
Purified (azide-free) anti-PSD95 [K28/74]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: WB, IHC, ICC; Size: 25 uL
Catalog No. 50-207-4016