Learn More
Biolegend Purified anti-rat CD326 (Ep-CAM)
Supplier:  Biolegend 251002
Encompass_Preferred
Purified anti-rat CD326 (Ep-CAM) [10D6]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: IHC-F; Size: 100 ug
Catalog No. 50-207-2582