Learn More
Biolegend Purified anti-S100A8
Supplier:  Biolegend 673451
Encompass_Preferred
Purified anti-S100A8 [A22124D]; Isotype: Mouse IgG2b, λ; Reactivity: Human; Apps: WB, IHC-P, ICC; Size: 25 μg
Catalog No. 50-286-5936