Learn More
Biolegend Purified anti-Serpin A12
Supplier:  Biolegend 624651
Encompass_Preferred
Purified anti-Serpin A12 [A16088B]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: IHC-P, WB; Size: 25 ug
Catalog No. 50-253-7757