Learn More
Biolegend Purified (azide-free) anti-SNAP-25
Supplier:  Biolegend 805002
Encompass_Preferred
Purified (azide-free) anti-SNAP-25 [SP12]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB, IP; Size: 25 uL
Catalog No. 50-207-3930