Learn More
Biolegend Purified anti-STAT1
Supplier:  Biolegend 661002
Encompass_Preferred
Purified anti-STAT1 [10C4B40]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IP; Size: 100 ug
Catalog No. 50-208-7182