Learn More
Biolegend Purified (azide-free) anti-Tau, 1-100
Supplier:  Biolegend 816603
Encompass_Preferred
Purified (azide-free) anti-Tau, 1-100 [43D]; Isotype: Mouse IgG1; Reactivity: Human; Apps: WB, IHC; Size: 25 uL
Catalog No. 50-207-4074