Learn More
Biolegend Purified anti-TICAM-2 (TRAM)
Supplier:  Biolegend 659402
Encompass_Preferred
Purified anti-TICAM-2 (TRAM) [9G1B30]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: WB; Size: 100 ug
Catalog No. 50-208-7162