Learn More
Biolegend Purified anti-UQCRC1
Catalog No. 502721758
Encompass_Preferred
Purified anti-UQCRC1 [O91G2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, ICC; Size: 100 μg
Catalog No. 50-272-1758 Supplier Biolegend Supplier No. 696202
Purified anti-UQCRC1 [O91G2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, ICC; Size: 100 μg