Learn More
Biolegend Purified anti-human CD66d/e
Supplier:  Biolegend 392802
Encompass_Preferred
Purified anti-human CD66d/e [308/3-3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IHC-P; Size: 100 ug
Catalog No. NC1648561