Learn More
Biolegend Purified anti-human CD8a
Supplier:  Biolegend 372902
Encompass_Preferred
Purified anti-human CD8a [C8/144B]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, Activ, FC, IHC-F, WB; Size: 100 ug
Catalog No. NC1541071