Learn More
Biolegend Purified anti-human CD279 (PD-1) (Maxpar Ready)
Supplier:  Biolegend 329941
Encompass_Preferred
Purified anti-human CD279 (PD-1) (Maxpar Ready) [EH12.2H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC, CyTOF; Size: 100 ug
Catalog No. NC1445689