Learn More
Biolegend Purified anti-human CD33
Supplier:  Biolegend 366602
Encompass_Preferred
Purified anti-human CD33 [P67.6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IHC-F; Size: 100 ug
Catalog No. NC1434581