Learn More
Biolegend Purified anti-human CD340 (erbB2/HER-2)
Supplier:  Biolegend 324402
Encompass_Preferred
Purified anti-human CD340 (erbB2/HER-2) [24D2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, ICC; Size: 100 ug
Catalog No. NC1462657