Learn More
iQ Biosciences Purified BALB/c Mouse CD3 Pan T Cells, 5M cells/vial
Catalog No. NC1985148
Encompass_Preferred
High-quality, high-viability (typically > 70%) mouse CD3 Pan T cells purified from spleen of BALB/c strain of mice. Highly enriched (> 90%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.
Catalog No. NC1985148 Supplier iQ Biosciences Supplier No. IQBM1T5
High-quality, high-viability (typically > 70%) mouse CD3 Pan T cells purified from spleen of BALB/c strain of mice. Highly enriched (> 90%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.