Learn More
iQ Biosciences Purified Beagle Canine CD14+ cells, 2M cells/vial
Catalog No. NC2331324
Encompass_Preferred
High-quality, high-viability (> 80%) CD14+ cells purified from beagle canine PBMCs by positive magnetic isolation. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2331324 Supplier iQ Biosciences Supplier No. IQBC1M2
High-quality, high-viability (> 80%) CD14+ cells purified from beagle canine PBMCs by positive magnetic isolation. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.