Learn More
Immunology Consultants Hamster (CHO) Lysosomal Phospholipase A2 - Recombinant, 0.025 mg
SDP
Supplier:  Immunology Consultants AG65012801Z
Encompass_Preferred
Recombinant CHO Lysosomal Phospholipase A2 (LPLA2; UniProt G3HKV9) is expressed in CHO‑S cells with a 6xHIS tag and purified by nickel affinity chromatography to >95% purity. Under reducing conditions, LPLA2 appears at ~56 kDa and is quantified using Bradford assay and A280. As a lysosomal enzyme involved in phospholipid metabolism, LPLA2 is a relevant CHO host cell protein that may co‑purify with biologics and impact product stability. This high‑quality recombinant antigen provides a dependable standard for ELISA development, antibody validation, and HCP profiling. Its CHO‑derived structure ensures biological relevance for bioprocess impurity studies, supporting accurate assay calibration and risk assessment. A strong choice for researchers evaluating lipid‑processing enzymes or refining downstream purification strategies.
Catalog No. 50-241-4274