Learn More
iQ Biosciences PURIFIED CYNO MONKEY CD4 10MIL
Catalog No. NC3452412
Encompass_Preferred
Purified Cynomolgus Macaque Monkey CD4 T Cells 10 million cells/vial. The cynomolgus macaque is the most commonly utilized non-human primate in biomedical research. According to some researchers, these monkeys are ideal models due to the 90-93% genetic similarity to and recent evolutionary divergence from humans. Purified cynomolgus CD4 T cells are a good source of cells to study the biology of CD4 T cells and their role in the immune system. They can be used for a variety of stimulation-dependent functional experiments to assess cytokine production and proliferation.
Catalog No. NC3452412 Supplier iQ Biosciences Supplier No. IQBMN1CD4T10
Purified Cynomolgus Macaque Monkey CD4 T Cells 10 million cells/vial. The cynomolgus macaque is the most commonly utilized non-human primate in biomedical research. According to some researchers, these monkeys are ideal models due to the 90-93% genetic similarity to and recent evolutionary divergence from humans. Purified cynomolgus CD4 T cells are a good source of cells to study the biology of CD4 T cells and their role in the immune system. They can be used for a variety of stimulation-dependent functional experiments to assess cytokine production and proliferation.