Learn More
Immunology Consultants Green Fluorescent Protein Control (GFP), 0.1 mg
SDP
Supplier:  Immunology Consultants AG450205Z
Encompass_Preferred
Recombinant Green Fluorescent Protein with a 6xHIS tag was expressed in E.coli and purified by affinity chromatography. Molecular weight is approximately 29 kDa. The protein is detected by Western blotting using anti-6HIS and anti-GFP antibodies. The proteinis determined to be >90% pure based on SDS-PAGE analysis.
Catalog No. 50-196-2150